Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARCKS Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735898 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5898 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5898 Supplier Proteintech Group Inc Supplier No. 510871AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Myristoylated alanine-rich protein kinase C substrate (MARCKS), also designated 80K or 80K-L, has been identified as a major cellular substrate for protein kinase C. Human MARCKS is a 332 amino acid protein with a calculated molecular weight of 31.534 kDa; however, it has been shown to run at 80-87 kDa on Western blot. The plasma membrane bound protein dissociates from the membrane upon phosphorylation by various PKC isoforms. In NIH/3T3 fibroblasts, PKC alpha and PKC epsilon, but not PKC delta, are responsible for MARCKS phosphorylation. MARCKS has been found to bind calmodulin, Actin and Synapsin and is a filamentous (F) Actin crosslinking protein.

Specifications

Antigen MARCKS
Applications Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MARCKS
Gene Accession No. P26645, P29966
Gene Alias 80K L, 80K L protein, MACS, MARCKS, PKCSL, PRKCSL
Gene Symbols Marcks
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17118, 4082
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.