Learn More
Invitrogen™ MARCO Monoclonal Antibody (PLK-1), PE, eBioscience™

Description
Description: This PLK-1 monoclonal antibody recognizes human MARCO, also known as SR-A6 or SCARA2. Applications Reported: This PLK-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This PLK-1 antibody has been pre-diluted and tested by flow cytometric analysis of human monocyte-derived macrophages. This may be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: μ8 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
Specifications
Specifications
| Antigen | MARCO |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | PLK-1 |
| Concentration | 5 μL/Test |
| Conjugate | PE |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | Marco |
| Gene Accession No. | Q9UEW3 |
| Gene Alias | AI323439; Ly112; macrophage receptor MARCO; Macrophage receptor with collagenous structure; MARCO; SCARA2; Scavenger receptor class A member 2; scavenger receptor class A, member 2; SR-A6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.