Learn More
Cayman Chemical MaresIn 2-d5 50ug
SDP
Supplier:  Cayman Chemical 2075750 UG
Encompass_Preferred
An active metabolite of tryptophan an antagonist of the NMDA and AMPA receptors as well as a7 nAChRs (EC50s 235 101 and 7 UM respectively) an agonist of AhR and GPR35 (EC50s 1 4 and 39 UM respectively) prevents lesioned hemisphere weight loss in a neonatal rat model of cerebral hypoxic-ischemia induced by carotid artery ligation at 300 mg/kg is protective against rhabdomere neurodegeneration in the eye in an Htt93Q transgenic Drosophila model of Huntingtons disease at 1 and 5 mg/ml cerebrospinal fluid levels are increased in patients with schizophrenia
Catalog No. 50-298-2155
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.