Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marginal Zone B Cells Mouse Anti-Rat, Brilliant Violet 711, Clone: HIS57, BD Optibuild™

Catalog No. BDB743345 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743345 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743345 Supplier BD Biosciences Supplier No. 743345
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HIS57 monoclonal antibody reacts with an unknown antigen that is highly expressed by most marginal zone B (MZ-B) cells in the spleen. In contrast, this antigen is weakly expressed, or not expressed at all, by other B-cell subpopulations. Rat MZ-B cells express low levels of CD45R (mAb HIS24) and sIgD and high levels of sIgM. The HIS57 mAb does not stain granulocytes and thymocytes. Immunohistochemical staining of normal spleen sections with HIS57 mAb produced a positive signal in the marginal zone and, to a lesser extent, in B-cell follicles. This marker can be used in combination with CD45R, sIgD, and sIgM t0 identify MZ-B cells in the rat.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Marginal Zone B Cells
Classification Monoclonal
Clone HIS57
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Splenic B Cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.