Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Marimastat, Matrix Metalloproteinase Inhibitor, Solid, 10 mg

Catalog No. 502706723
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6723 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6723 Supplier Abcam Supplier No. AB14127610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Broad-spectrum matrix metalloproteinase inhibitor with >98% purity.

  • Purity: >98%
  • Molecular Weight: 331.41 Da
  • Form: Solid
  • Activity: Potent, broad-spectrum matrix metalloproteinase (MMP) inhibitor
  • Applications: Prevents or reduces spread and growth of tumors
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Matrix Metalloproteinases
Buffer 100% Marimastat
Form Solid
Shipping Condition Ambient
Content And Storage Store at -20°C.
Quantity 10 mg
For Use With (Application) Matrix metalloproteinase inhibition, Cancer research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.