Learn More
Grainger Marine Eye Fitting 316 SS 1/16 in dia.
Supplier:  Grainger 16X739
Encompass_Preferred
The LOOS Marine Eye Fitting is a 316 stainless steel marine eye fitting with a 1/16 inch NPT female pipe thread. It has a working pressure of 3,000 psi and temperature range of -20 to 250 degrees Fahrenheit. This fitting allows for a fixed or swivel attachment point and is designed for use in marine environments. With its high strength and corrosion resistance, it is well suited for securing lines, wires, and cables on boats. The compact size makes it easy to install while still providing a secure hold.
Catalog No. 50-276-5697
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.