Learn More
MARK2 Polyclonal Antibody

Description
MARK2 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | MARK2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 4 μg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 |
| Gene | MARK2 |
| Gene Accession No. | O08679, Q05512, Q7KZI7 |
| Gene Alias | ELKL motif kinase; ELKL motif kinase 1; Emk; EMK1; EMK-1; MAP/microtubule affinity regulating kinase 2; MAP/microtubule affinity-regulating kinase 2; MARK2; mark2 {ECO:0000312; MGC99619; microtubule affinity regulating kinase 2; OTTHUMP00000218338; OTTHUMP00000218339; OTTHUMP00000218340; OTTHUMP00000218343; Par-1; PAR1 homolog; PAR1 homolog b; Par1b; Par-1b; RGD:708483}; Ser/Thr protein kinase PAR-1B; serine/threonine kinase; serine/threonine protein kinase EMK; serine/threonine-protein kinase MARK2; testicular tissue protein Li 117 |
| Gene Symbols | MARK2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.