Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MARK4 Recombinant Protein

Catalog No. 89968698 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-968-698 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-968-698 Supplier Abnova™ Supplier No. H00057787P01
Add to Cart
Add to Cart

Used for ELISA, Western Blotting (Recombinant Protein), Antibody Production and Protein Array

Sequence: MRATPLAAPAGSLSRKKRLELDDNLDTERPVQKRARSGP QPRLPPCLLPLSPPTAPDRATAVATASRLGPYVLLEPEE GGRAYRALHCPTGTEYTCKVYPVQEALAVLEPYARLPPH KHVARPTEVLAGTQLLYAFFTRTHGDMHSLVRSRHRIPE PEAAVLFRQMATALAHCHQHGLVLRDLKLCRFVFADRER KKLVLENLEDSCVLTGPDDSLWDKHACPAYVGPEILSSR ASYSGKAADVWSLGVALFTMLAGHYPFQDSEPVLLFGKI RRGAYALPAGLSAPARCLVRCLLRREPAERLTATGILLH PWLRQDPMPLAPTRSHLWEAAQVVPDGLGLDEAREEEGD REVVLYG

Specifications

Accession Number AAH09049
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50 mM Tris HCl, 10 mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 57787
Molecular Weight (g/mol) 34.43 kDa
Name MARK4 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 2 μg
Source Wheat Germ (in vitro)
Immunogen MAALRQATAAARCRCRQPQPFLLACLHGGAGGPEPLSHFEVEVCQLPRPGLRGVLFRRVAGTALAFRTLVTRISNDLEL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias FLJ90097, KIAA1860, MARKL1, Nbla00650
Common Name MARK4
Gene Symbol MARK4
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.