Learn More
Sartorius MARKER CARTRIDGE SINGLE
Supplier:  Sartorius 97018
Encompass
Volume37 mL sufficient to run 100 384-well plates. Run time is dependent on protocol.StorageStore at Room Temperature.NotesEnhanced Flush Cartridges are only compatible on iQue3 systems running ForeCyt versions 7.1 or higher. Cartridge expires 30 days after opening.
Catalog No. 50-289-1734