Learn More
Sartorius MARKER CARTRIDGE SINGLE
Catalog No. 502891612 Shop All Sartorius Products
Encompass
Volume37 mL sufficient to run 100 384-well plates.Run time is dependent on protocol.StorageStore at Room Temperature. Once opened store at 2-8°C. Use Enhanced Marker Cartridges enable the use of the marker experiments option in ForeCyt. In “Marker Experiment” mode the sample probe travels to the marker cartridge and aspirates a small quantity of a fluorescent bead solution between each sample. The between-well beads are utilized by the software to perform unassisted well identification.NotesMarker Cartridges are only compatible on iQue3 systems running ForeCyt versions 7.1 or higher. Cartridge expires 30 days after opening.
Catalog No. 50-289-1612 Supplier Sartorius Supplier No. 97026
Volume37 mL sufficient to run 100 384-well plates.Run time is dependent on protocol.StorageStore at Room Temperature. Once opened store at 2-8°C. Use Enhanced Marker Cartridges enable the use of the marker experiments option in ForeCyt. In “Marker Experiment” mode the sample probe travels to the marker cartridge and aspirates a small quantity of a fluorescent bead solution between each sample. The between-well beads are utilized by the software to perform unassisted well identification.NotesMarker Cartridges are only compatible on iQue3 systems running ForeCyt versions 7.1 or higher. Cartridge expires 30 days after opening.