Learn More
Marketlab, Inc. Marketlab Tube Caps
Description
- Will not aerosol when opened
- Designed to hold under centrifugation, agitation, refrigeration, and freezing
- Stretchable, over-locking design prevents evaporation
- Easy removal and application with only one hand
Specifications
Specifications
| Product Type | Screw Cap |
| Autoclavable | Not Autoclavable |
| Material | Polyethylene |
| Color | Blue |
| Sterility | Non-sterile |
| Disposable | Reusable |