Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Marking Flags

Catalog No. 19103885 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-103-885 Blue
19-103-882 Orange
19-103-884 Yellow
19-103-886 White
19-103-883 Red
19-103-887 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-103-885 Supplier Brady™ Supplier No. 98175
Add to Cart
Add to Cart

Ideal for ground marking to easily identify buried lines and cable

  • Colorful Marking Flags help identify possible hazards buried underground
  • Custom legends are also available
  • Flags attach to a 30 in. steel rod

Specifications

Color Blue
Width (English) 4 in.
Width (Metric) 1676.4 mm
Packaging Quantity 100 Pk.
Type Marking Flag
Height (English) 5 in.
Length (English) 30 in.
Material Steel, Plastic
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.