Learn More
Description
- Colorful Marking Flags help identify possible hazards buried underground
- Custom legends are also available
- Flags attach to a 30 in. steel rod
Specifications
Specifications
| Color | White |
| Height (English) | 5 in. |
| Length (English) | 30 in. |
| Packaging Quantity | 100 Pk. |
| Width (English) | 4 in. |
| Width (Metric) | 1422.4 mm |
| Material | Steel, Plastic |
| Type | Marking Flag |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.