Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Marking Tape Roll: Adhesive Vinyl, Solid Color, Green

Catalog No. 19103843 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-103-843 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-843 Supplier Brady™ Supplier No. 121125
Add to Cart
Add to Cart

Permanent acrylic adhesive sticks to a wide variety of surfaces

  • Indoor/outdoor vinyl film withstands exposure to moisture, chemicals and handling.
  • Up to 3 year average outdoor durability

Specifications

Color Green
Packaging Quantity 50 ft./Roll
Width (Metric) 78.74 mm
Certifications/Compliance ASTM D 2979
Displayed Text Blank
Length (English) 50 ft.
Adhesive Type Permanent Acrylic
Material Vinyl
Type Marking Tape
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.