Description
- Eight different gauge nichrome wires of equal length
- Scale on mounting board for easy measurements
Specifications
Specifications
| For Use With (Application) | showing resistivity of different gauge wires |
| Includes | Eight different gauge Nichrome wires of equal length, scale on mounting board |
| Product Type | Resistance Board Set |