Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARVELD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109941
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109941 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109941 Supplier Invitrogen™ Supplier No. PA5109941
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLPPPPRQPP PQARAARGAV RLQRPFLRSP LGVLRLLQLL AGAAFWITIA.

MARVELD1, (MARVEL domain-containing 1), is one of the MARVEL (MAL and related proteins for vesicle trafficking and membrane link) domain-containing proteins. It is a nuclear-localized protein that is widely expressed in normal human tissues. While it has been reported to be overexpressed in colon cancer, MARVELD1 protein levels are generally lower in several primary tumors including ovary, breast, kidney, bladder and liver. This lower level of MARVELD1 expression in tumors is thought to be due to a higher methylation status of the MARVELD1 promoter as treatment of MDA-MB-361 with 5-aza-2'-deoxycytidine restores MARVELD1 expression, suggesting that loss of MARVELD1 expression may contribute to tumorgenicity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARVELD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MARVELD1
Gene Accession No. Q7TQJ1, Q9BSK0
Gene Alias AI504298; bA548K23.8; GB14; MARVD1; MARVEL (membrane-associating) domain containing 1; MARVEL domain containing 1; MARVEL domain-containing protein 1; Marveld1; MRVLDC1; putative MARVEL domain-containing protein 1; RP11-548K23.8
Gene Symbols MARVELD1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-50 of human MARVELD1 (NP_1136721).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 277010, 309375, 83742
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.