Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARVELD3 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735013 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5013 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5013 Supplier Proteintech Group Inc Supplier No. 256671AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function remains unknown.

Specifications

Antigen MARVELD3
Applications Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MARVELD3
Gene Accession No. Q96A59
Gene Alias MARVD3, MARVEL domain containing 3, MARVELD3, MRVLDC3
Gene Symbols MARVELD3
Host Species Rabbit
Immunogen MARVELD3 Fusion Protein Ag22492
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91862
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.