Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mas Antibody Blocking Peptide
SDP

Catalog No. NB398512 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398512 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398512 Supplier Novus Biologicals™ Supplier No. NBP178444PEP
Add to Cart
Add to Cart

Blocking Peptide

Specifications

Gene ID (Entrez) 4142
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
For Use With (Application) This peptide is useful as a blocking peptide for NBP1-78444. For further blocking peptide related protocol, click here .
Gene Alias MAS, MAS1 oncogene, MGC119966, proto-oncogene Mas
Gene Symbol MAS1
Quantity 0.1 mg
Research Category Cancer, GPCR, Neuroscience
Storage Buffer Peptide dissolved in dH20. Contains no BSA.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.