Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAS™ Diabetes Control and MAS Diabetes Max Control

Catalog No. 21124393 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Multi-pack
Vista Vials
Level:
2
Bi-level
Quantity:
6 x 1 mL
6 x 2 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Level Quantity
21-124-393 Multi-pack 2 6 x 2 mL
23-640-25 Multi-pack Bi-level 6 x 1 mL
23-640-27 Vista Vials Bi-level 6 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 21-124-393 Supplier Thermo Scientific™ Supplier No. B40000687

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Monitor hemoglobin A1c methods with Thermo Scientific™ MAS™ Diabetes Control, a bi-level control available in a convenient Vista™ vial format.

Monitor hemoglobin A1c methods with bi-level Thermo Scientific™ MAS™ Diabetes and MAS Diabetes Max Controls. These liquid, ready-to-use, assayed diabetes controls have been assayed for most major instrument systems. The MAS Diabetes MAX controls are packaged in convenient Load-and-Go tubes with barcodes1 provided for most major platforms, simplifying the daily QC routine and saving significant amounts of time.

  • Liquid, ready-to-use whole blood matrix to better mimic patient sample matrix
  • Load-and-Go barcoded tubes for improved efficiency and daily time savings
  • Human whole blood matrix to better mimic patient samples
  • Monitoring of hemoglobin A1c testing on major platforms
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 6 x 2 mL, frozen
Description Multi-pack
Concentration Value assignments available on Lablink xL
Detectable Analytes HbA1c
Detection Method (Not applicable as it is a QC)
Format Load-and-Go tubes
For Use With (Equipment) Instrument agnostic
Sample Type Whole Blood
Type QC
For Use With (Application) MAS Diabetes is intended for use as an assayed control material for monitoring Glycosylated Hemoglobin (A1c) assay procedures.
Intended Use MAS Diabetes is intended for use as an assayed control material for monitoring Glycosylated Hemoglobin (A1c) assay procedures.
Level 2
Quantity 6 x 2 mL
Source Human matrix
Stability
  • 60 days unopened stability at 2 to 8°C
  • 30 days open vial stability at 2 to 8°C
Form Frozen Liquid
Show More Show Less

1Barcodes available for selected instruments.

Not all products are CE marked or have FDA clearance for sale in the U.S. Availability of products in each country depends on local regulatory marketing authorization status.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.