Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAS™ Diabetes Control and MAS Diabetes Max Control
GSA_VA

Catalog No. 2364025 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Multi-pack
Vista Vials
Level:
2
Bi-level
Quantity:
6 x 1 mL
6 x 2 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Level Quantity
23-640-25 Multi-pack Bi-level 6 x 1 mL
21-124-393 Multi-pack 2 6 x 2 mL
23-640-27 Vista Vials Bi-level 6 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-640-25 Supplier Thermo Scientific™ Supplier No. DBCLMP

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Monitor hemoglobin A1c methods with Thermo Scientific™ MAS™ Diabetes Control, a bi-level control available in a convenient Vista™ vial format.

Monitor hemoglobin A1c methods with bi-level Thermo Scientific™ MAS™ Diabetes and MAS Diabetes Max Controls. These liquid, ready-to-use, assayed diabetes controls have been assayed for most major instrument systems. The MAS Diabetes MAX controls are packaged in convenient Load-and-Go tubes with barcodes1 provided for most major platforms, simplifying the daily QC routine and saving significant amounts of time.

  • Human whole blood matrix
  • Assayed values available for most major analyzers

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Two year shelf life at -25° to -15°C; 30 days open vial at 2° to 8°C
Description Multi-pack
Detectable Analytes Hemoglobin A1c
Sample Type Whole Blood
Type Control
Level Bi-level
Packaging 6 Pk.
Quantity 6 x 1 mL

1Barcodes available for selected instruments.

Not all products are CE marked or have FDA clearance for sale in the U.S. Availability of products in each country depends on local regulatory marketing authorization status.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.