Learn More
Thermo Scientific™ MAS™ Immunology Controls
GSA_VA
Monitor the performance of various serum protein and serology test procedures with ready-to-use, liquid assayed multi-analyte immunology controls.
Supplier:  Thermo Scientific™ IC101
 View more versions of this product
Catalog No. 23-100004
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Monitor the performance of various serum protein and serology test procedures with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ Immunology Controls. These controls are also available in a convenient Vista™ vial format.
  • Assayed for most major instrument systems
  • Three levels provide monitoring of the full assay range
  • Contains specific serum proteins, including Antistreptolysin, CRP, Prealbumin and Rheumatoid Factor
  • Human-serum matrix
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions