Learn More
Thermo Scientific™ MAS™ Immunology Controls
Monitor the performance of various serum protein and serology test procedures with ready-to-use, liquid assayed multi-analyte immunology controls.
| Catalog Number | Mfr. No. | Description | dimLevel | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Description | dimLevel | Price | Quantity & Availability | ||||
23-100004
![]() |
Thermo Scientific™
IC101 |
N/A | N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-100005
![]() |
Thermo Scientific™
IC202 |
N/A | N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-100006
![]() |
Thermo Scientific™
IC303 |
N/A | N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
|
23-100-131
|
Thermo Scientific™
ICV1 |
N/A | N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
|
23-100-132
|
Thermo Scientific™
ICV2 |
N/A | N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
|
23-100-133
|
Thermo Scientific™
ICV3 |
N/A | N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
Description
Monitor the performance of various serum protein and serology test procedures with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ Immunology Controls. These controls are also available in a convenient Vista™ vial format.
- Assayed for most major instrument systems
- Three levels provide monitoring of the full assay range
- Contains specific serum proteins, including Antistreptolysin, CRP, Prealbumin and Rheumatoid Factor
- Human-serum matrix
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.