Learn More
Thermo Scientific™ MAS™ Immunology Controls
Monitor the performance of various serum protein and serology test procedures with ready-to-use, liquid assayed multi-analyte immunology controls.
Supplier: Thermo Scientific™ IC303
Description
Monitor the performance of various serum protein and serology test procedures with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ Immunology Controls. These controls are also available in a convenient Vista™ vial format.
- Assayed for most major instrument systems
- Three levels provide monitoring of the full assay range
- Contains specific serum proteins, including Antistreptolysin, CRP, Prealbumin and Rheumatoid Factor
- Human-serum matrix