Learn More
Thermo Scientific™ MAS™ Immunology Controls
Monitor the performance of various serum protein and serology test procedures with ready-to-use, liquid assayed multi-analyte immunology controls.
Supplier:  Thermo Scientific™ ICV2
 View more versions of this product
Catalog No. 23-100-132
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Monitor the performance of various serum protein and serology test procedures with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ Immunology Controls. These controls are also available in a convenient Vista™ vial format.
  • Assayed for most major instrument systems
  • Three levels provide monitoring of the full assay range
  • Contains specific serum proteins, including Antistreptolysin, CRP, Prealbumin and Rheumatoid Factor
  • Human-serum matrix
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions