Learn More
Description
Consolidate general chemistry and serum protein QC processing into a single product offering three distinct levels with Thermo Scientific™ MAS™ Omni·CORE Controls. Value assignment is provided for key integrated instrument systems.
- Three distinct levels
- Liquid, ready-to-use
- Human-serum based matrix
- Assayed values available for most analyzers
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Omni CORE Control, Assayed |
| Detectable Analytes | Acetaminophen, Albumin, Alkaline Phosphatase, alpha-1-Acid Glycoprotein, alpha-1-Antitrypsin, alpha-2-Macroglobulin, ALT, Amikacin, Amylase, Amylase (Pancreatic), Antistreptolysin O (ASO), Apolipoprotein A1, Apolipoprotein B, AST, beta-2-microglobulin (B2M), Bile Acids, Bilirubin Conjugated, Bilirubin Direct (DBIL), Bilirubin Neonatal, Bilirubin Total (TBIL), BUN, C3 Complement, C4 Complement, Caffeine, Calcium, Carbamazepine, Ceruloplasmin, Chloride, Cholesterol, Creatine Kinase (CK), CO2, Copper, Cortisol, C-Reactive Protein (CRP), Creatinine, Digoxin, Disopyramide, Ethanol, Ethosuximide, Ferritin, Gentamicin, GGT, Glucose, Glutamate Dehydrogenase (GLDH), Haptoglobin, HDL, IgA, IgE, IgG, IgM, Iron, Iron Binding Capacity (IBC), Iron Binding Capacity, Total (TIBC), Iron Binding Capacity, Unsaturated (UIBC), Lactic Acid, LDH, LDL, Lidocaine, Lipase, Lipoprotein (a), Lithium, Magnesium, Methotrexate, N-acetylprocainamide (NAPA), Osmolality, Phenobarbital, Phenytoin, Phospholipids, Phosphorus, Potassium, Prealbumin, Primidone, Procainamide, Protein Electrophoresis, Pseudocholinesterase (PCHE), Quinidine, Rheumatoid Factor, Salicylate (SALY), Sodium, Thyroxine (T4 Total), Theophylline, Tobramycin, Total Protein, Transferrin (TRF), Tricyclic Antidepressants, Triglycerides, Thyroid Stimulating Hormone (TSH), T-uptake, Uric Acid, Valproic Acid, Vancomycin, Zinc |
| Sample Type | Serum |
| Type | Control |
| Level | 3 |
| Packaging | 6 Pk. |
| Quantity | 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.