Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAS™ Omni·IMMUNE™ PRO Immunoassay Controls
GSA_VA

Catalog No. 22640048 Shop All Thermo Scientific Products
Change view
Click to view available options
Level:
1
2
3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
22-640-048 3
22-640-046 1
22-640-047 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-640-048 Supplier Thermo Scientific™ Supplier No. OPRO303

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Consolidate your immunoassay and tumor marker controls into a single QC product with MAS Omni·IMMUNE PRO Immunoassay Controls.

Consolidate your immunoassay and tumor marker controls into a single QC product with Thermo Scientific™ MAS™ Omni·IMMUNE™ PRO Immunoassay Controls. This QC product contains the same analyte panel as the Omni·IMMUNE control but also has the following additional analytes: Anti-Thyroglobulin (Anti-Tg), Anti-Thyroid Peroxidase (Anti-TPO) and Sex Hormone Binding Globulin (SHBG).

  • Liquid, ready-to-use
  • Human-based serum matrix
  • Value assignment provided for most major analyzers

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -25°C to -15°C
Description Omni IMMUNE PRO Control, Assayed
Detectable Analytes 17-OH-Progesterone, 25-OH-Vitamin D, Acetaminophen, Adrenocorticotropic Hormone (ACTH), Aldosterone, alpha-Fetoprotein (AFP), Amikacin, Anti-Tg, Anti-TPO, Benzodiazepines, Oxazepam, β-hCG, CA-125, CA 15-3, CA 19-9, Carbamazepine, Carcinoembryonic Antigen (CEA), Cortisol, C-peptide, DHEA Sulfate, Digoxin, Disopyramide, Estradiol, Estriol (Free), Ethosuximide, Ferritin, Folate (Vitamin B9), Fructosamine, Follicle Stimulating Hormone (FSH), Gastrin, Gentamicin, Growth Hormone, hCG, Homocysteine, IgE, Insulin-like Growth Factor 1 (IGF-1), Insulin, Luteinizing Hormone (LH), Lidocaine, N-acetylprocainamide (NAPA), Prostatic Acid Phosphatase (PAP), Phenobarbital, Phenytoin, Phenytoin (Free), Primidone, Procainamide, Procalcitonin (PCT), Progesterone, Prolactin, PSA (Free), PSA (Total), Parathyroid Hormone (PTH), Quinidine, Salicylate (SALY), Sex Hormone-binding Globulin (SHBG), T3 (Free), Triiodothyroxine (T3 Total), T4 (Free), Thyroxine (T4 Total), Thyroxine-binding Globulin (TBG), Testosterone, Theophylline, Thyroglobulin, Tobramycin, Tricyclic Antidepressants, Thyroid Stimulating Hormone (TSH), T-uptake, Valproic Acid, Valproic Acid (Free), Vancomycin, Vitamin B12
Sample Type Serum
Type Control
Level 3
Packaging 6 Pk.
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.