Learn More
Thermo Scientific™ MAS™ Omni·Infectious™ Quality Controls
Catalog No. 10027480 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
MAS HIV p24 Positive Control
MAS Infectious Negative Control
MAS Omni Infectious BSI Positive Control Panel
MAS Syphilis Positive Control
MAS anti-HBs Positive Control
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
10-027-480 MAS Omni Infectious BSI Positive Control Panel
10-027-481 MAS HIV p24 Positive Control
10-027-482 MAS anti-HBs Positive Control
10-027-483 MAS Syphilis Positive Control
10-027-484 MAS Infectious Negative Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 10-027-480 Supplier Thermo Scientific™ Supplier No. 10027480
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Assess the performance of Serological assays for viral and bacterial pathogens using the Thermo Scientific MAS Omni Infectious Quality Controls.
Assess the performance of serological assays for viral blood stream infections using the Thermo Scientific MAS Omni Infectious Quality Controls. These third-party control panels include individual positive controls for anti-HBs, HIV-p24 and Syphilis as well as consolidated HIV, HCV, HBV and HTLV for efficiency.
  • 30 days open vial stability for Syphilis; anti-HBs and HIV p24 at 2 to 8°C
  • Ready to use straight from the refrigerator
  • Human serum matrix
  • BSI Positive and Infectious Negative
  • BSI Positive has Multiple analytes in a single vial
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Description MAS Omni Infectious BSI Positive Control Panel
Detectable Analytes HBsAg, anti-HIV, anti-HTLV, anti-HBc, anti-HCV, anti-HBs
Type Control
Packaging One box of control contains five tubes of positive control
Quantity 5 x 5.0 mL