Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAS™ UA Controls
GSA_VA

Catalog No. 23038384 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
UA Controls, Assayed
Level:
1
2 (abnormal)
Quantity:
15 mL
60 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Level Quantity
23-038-384 UA Controls, Assayed 1 60 mL
23-038-381 UA Controls, Assayed 2 (abnormal) 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-038-384 Supplier Thermo Scientific™ Supplier No. UAB160

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.

This item is not returnable. View return policy

Monitor urine dipstick and microscopic components with bi-level controls with improved room temperature storage capability.

Monitor urine dipstick and microscopic components with Thermo Scientific™ MAS™ UA Controls, bi-level controls with improved room temperature storage capability. Liquid and ready-to-use, this urine control is assayed for most major reagents strips and strip systems. It includes 10 commonly-assayed dipstick tests plus Microalbumin, Creatinine and hCG.

Ordering Information:
Available in convenient 15mL dropper bottles

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 24 month shelf life at 2° to 8°C; Open Vial: Refrigerated 2° to 8°C for 3 months; Room Temperature 18° to 25°C for 6 weeks
Description UA Controls, Assayed
Detectable Analytes Bilirubin, Blood, Creatinine, Crystals, Glucose, hCG, Ketones, Leukocyte Esterase, Microalbumin, Nitrite, pH, Potassium, Protein, Red Blood Cells, Sodium, Specific Gravity, Urobilinogen, White Blood Cells
Sample Type Urine
Type Control
Level 1
Packaging 4 Pk.
Quantity 60 mL
Stability 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.