Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAS™ UA DipTube Control
GSA_VA

Catalog No. UATMP Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
12 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
UATMP 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. UATMP Supplier Thermo Scientific™ Supplier No. UATMP

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simplify QC during urinalysis testing with room-temperature, stable Thermo Scientific™ MAS™ UA DipTube Control, conveniently packed for time and cost savings.

Simplify the process of quality control during urinalysis testing with room-temperature, stable Thermo Scientific™ MAS™ UA DipTube Control. This liquid assayed urinalysis control for procedures used in routine urinalysis testing is conveniently-packed to afford immediate time and cost savings.

Features:
  • Key urinalysis test parameters provided in a single control
  • Tube capacity common within urinalysis test environment
  • Assayed for most major reagent strips and strip systems
  • Human urine matrix

Ordering Information:

Packaged in 12mL dip tube format; 5 vials of each level (1 and 2)

Requires:

  • Slides
  • Pipetters
  • Centrifuge capable of generating an RCF appropriate for the microscopic method used by the laboratory
  • Microscope with a 10x objective and a 40x objective lens
  • Chemical reagent strips

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Assayed, Bi-level Multi-Pack; 10 x 12mL
Detectable Analytes Bilirubin, Blood, Creatinine, Crystals, Glucose, hCG, Ketones, Leukocyte Esterase, Microalbumin, Nitrite, pH, Potassium, Protein, Red Blood Cells, Sodium, Specific Gravity, Urobilinogen, White Blood Cells
Sample Type Urine
Type Control
Level Bi-level
Packaging 10 Pk.
Quantity 12 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.