Learn More
Description
Monitor the most commonly tested urine chemistry analytes with liquid, assayed Thermo Scientific™ MAS™ UrichemTRAK Controls, available in a convenient Vista™ vial format. Ready-to-use and requiring no additional preparation, these controls will not interfere with ISEs.
- Liquid, human urine matrix
- Open and use; no equilibration required
- Two-year open-vial stability
- Individual levels or multi-pack configurations
- Stable for 24 months at 2° to 8°C
- Assayed values available for most major analyzers
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | UrichemTRAK Controls, Assayed |
| Detectable Analytes | Amylase, Calcium, Chloride, Cortisol, Creatinine, Glucose, hCG, Magnesium, Microalbumin, Osmolality, pH, Phosphorus, Potassium, Sodium, Specific Gravity, Total Protein, Urea Nitrogen, Uric Acid |
| Sample Type | Urine |
| Type | Control |
| Level | 2 |
| Packaging | 6 pk. |
| Quantity | 15 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
