Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAS™ Urinalysis Controls
GSA_VA

Catalog No. 23030127 Shop All Thermo Scientific Products
Change view
Click to view available options
Level:
1
3
Quantity:
15 mL
60 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level Quantity
23-030127 3 15 mL
23-030125 1 15 mL
23-030128 1 60 mL
23-030131 3 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-030127 Supplier Thermo Scientific™ Supplier No. 024224

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.

This item is not returnable. View return policy

Combine dipstick methods and microscopic components with bi-level, liquid assayed MAS Urinalysis Controls, assayed for most reagent strips and strip systems.

Combine dipstick methods and microscopic components with bi-level, liquid assayed Thermo Scientific™ MAS™ Urinalysis Controls. This control is assayed for most major reagent strips and strip systems.

  • Human urine matrix

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Urinalysis Control, Assayed
Detectable Analytes Bilirubin, Blood, Creatinine, Crystals, Glucose, hCG, Ketones, Leukocyte Esterase, Microalbumin, Nitrite, Osmolality, pH, Potassium, Protein, Red Blood Cells, Sodium, Specific Gravity, Urobilinogen, White Blood Cells
Sample Type Urine
Type Control
Level 3
Packaging 4 Pk.
Quantity 15 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.