Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MASH1 Monoclonal Antibody (24B72D11), eBioscience™, Invitrogen™

Catalog No. 502550884
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-255-0884 25 μg
50-112-2694 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-255-0884 Supplier Invitrogen™ Supplier No. 14579480
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The monoclonal antibody 24B72D11 recognizes mouse and rat Mammalian achaete-scute homolog 1 (MASH1), a 34 kDa basic helix-loop-helix (bHLH) transcription factor. MASH1, like family members Olig2 and Ngn2, is expressed in progenitor cells throughout the nervous system and retina and is a key regulator of cell fate specification. Expression of MASH1 is regulated by the Delta/Notch1 pathway through direct binding of Hes1 to MASH1 as well as competitive binding to MASH1 hetero-dimerizing proteins. At a transcriptional level, MASH1 expression is controlled by notch-mediated regulation of sox2, which binds to the MASH1 promoter region up-regulating expression. MASH1 has been implicated in the formation of multiple neural cell types including dopaminergic, serotonergic, glutaminergic and GABAergic neurons. Overexpression of MASH1 has been shown in small cell lung carcinoma and medullary thyroid cancer. Applications Reported: This 24B72D11 antibody has been reported for use in western blotting, immunohistochemical staining of frozen tissue sections, and immunocytochemistry. Applications Tested: This 24B72D11 antibody has been tested by western blot on reduced PC12 cells and can be used at less than or equal to 5 μg/mL. The 24B72D11 antibody has also been tested by immunocytochemistry on fixed and permeablized PC12 cells at can be used at less than or equal to 20 μg/mL.

ASCL1 (also known as ASH1) is a basic helix-loop-helix transcription factor that is required for early development of the nervous system. Expressed in fetal brain, ASCL1 is essential for the proper development of autonomic neurons and for the survival of subsets of autonomic neurons. ASCL1 interaction with MEF-2A may regulate the expression of specific genes that are critical for the formation of distinct neuronal circuits within the central nervous system. The high level of ASCL1 expression in neuroendocrine tumors, such as medullary thyroid cancer, small cell lung cancer and lung cancer with neuroendocrine features may provide a useful marker for cancers with neuroendocrine features. Mapping to human chromosome 12, the ASCL1 gene contains a trinucleotide repeat region, making this locus a candidate for inherited disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MASH1
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 24B72D11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Ascl1
Gene Accession No. P19359, Q02067
Gene Alias achaete scute protein; achaete-scute complex homolog 1; achaete-scute complex homolog 1 (Drosophila); achaete-scute complex homolog-like 1; achaete-scute complex-like 1; achaete-scute complex-like protein 1; achaete-scute family bHLH transcription factor 1; achaete-scute homolog 1; AI225900; Ascl1; ASH1; ASH-1; bHLHa46; Class A basic helix-loop-helix protein 46; HASH1; KMT2H; mammalian achaete scute homolog 1; Mash1; mASH-1
Gene Symbols Ascl1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17172, 64186
Target Species Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.