Learn More
Selleck Chemical LLC Masitinib (AB1010) 10mM/1mL 790299-79-5
SDP
Supplier:  Selleck Chemical LLC S106410MM/1ML
Encompass_Preferred
Masitinib is a novel inhibitor for Kit (c-Kit) and PDGFR?/? with IC50 of 200 nM and 540 nM/800 nM, weak inhibition to ABL and c-Fms. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0883
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.