Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Masking Tape 2307

Catalog No. 19072119 Shop All 3M Products
Change view
Click to view available options
Quantity:
24 Rolls/Cs.
36 Rolls/Cs.
Dimensions (L x W):
55 m x 24 mm
55 x 36 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Dimensions (L x W)
19-072-119 24 Rolls/Cs. 55 x 36 mm
19-072-024 36 Rolls/Cs. 55 m x 24 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-072-119 Supplier 3M™ Supplier No. 7000123532
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Masking Tape 2307 was formulated for general use, with a broad tack that will adhere to a variety of surfaces. The rubber adhesive sticks instantly with moderate strength. The crepe paper backing conforms over irregular surfaces and around corners while holding reliably. It is especially useful in woodworking and cabinet making applications.

  • Easily identifiable, general purpose tape for woodworking
  • Rubber adhesive offers broad tack and moderate adhesion
  • Sticks quickly and holds reliably
  • Crepe-paper backing conforms over irregular surfaces, contours, and around corners
  • Rubber adhesive sticks instantly with moderate strength.
  • Crepe paper backing conforms over irregular surfaces and around corners while holding reliably.
  • Especially useful in woodworking and cabinet making applications.
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
Material Rubber
Product Type Masking Tape
For Use With (Application) Automotive, Design and Construction, Manufacturing, Mining, Oil and Gas, Transportation
Model 2307
Quantity 24 Rolls/Cs.
Thickness (English) 5.2 mil
Dimensions (L x W) 55 x 36 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.