Learn More
Vernier™ Mass for Dynamics Cart
SDPSCI_ED
Catalog No. S16194ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Dynamics System Cart
Catalog No. For Use With (Equipment)
S16194ND Dynamics System Cart
1 options
Catalog No. S16194ND Supplier Vernier™ Supplier No. MASS
Add to Cart
Add to Cart
Includes
Mass for Dynamics Cart
This 500 g mass is used to add mass to a cart for dynamics experiments. It includes hardware for attaching to a Vernier Dynamics System cart or a PASCO dynamics cart, and leaves room for both a Dual-Range Force Sensor and an Accelerometer. Not compatible with newer plastic dynamics carts.
  • This 500 g mass is used to add mass to a metal dynamics cart for dynamics experiments.
  • It includes hardware for attaching to a metal Dynamics System cart or a PASCO dynamics cart, and leaves room for both a Dual-Range Force Sensor and an Accelerometer.
  • It is not compatible with the plastic carts included with the Dynamics Cart and Track System family of products.
Warranty and Services
5-year limited warranty
Specifications
Description 500 g mass
Includes Hardware for attaching
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) Dynamics System Cart