Learn More
REFEYN INC MassGlass KV - Sample Prep Kit, to enable 140 sample measurements.
SDP
Supplier:  REFEYN INC MPCON21026
Encompass
"Sample carrier preparation kit to enable 140 sample measurements. Compatible with KaritroMP. This includes:- 10 ready-to-use coated MassGlass KV carrier slides (10 slides/box, 3 months shelf life)- 10 sample carrier lids- 10 sample well cassettes (14 sample wells/cassette)- 1 box of lens Cleaning Tissues (25 sheets)"
Catalog No. NC3737108