Learn More
StatLab MASSONS TRICHROME STAIN KIT
SDP
Supplier:  StatLab KTMTRPT
Encompass_Preferred
This kit stains for collagen and muscle. Kit contains Bouins Fluid Pint, Bieberich Scarlet/Acid Fucshin Pint, Phosphomolybdic/Phoshotungstic Acid Pint, Aniline Blue Stain Pint, 1% Acetic Acid Pint, and Weigert's A & B Hematoxylin Pint each.
Catalog No. NC9900705
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.