Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mast Cell Chymase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579053
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579053 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579053 Supplier Invitrogen™ Supplier No. PA579053
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human PC-3 whole cell, human HepG2 whole cell, rat liver tissue, mouse HEPA1-6 whole cell. IHC: human placenta tissue.

This gene product is a chymotryptic serine proteinase that belongs to the peptidase family S1. It is expressed in mast cells and thought to function in the degradation of the extracellular matrix, the regulation of submucosal gland secretion, and the generation of vasoactive peptides. In the heart and blood vessels, this protein, rather than angiotensin converting enzyme, is largely responsible for converting angiotensin I to the vasoactive peptide angiotensin II. Angiotensin II has been implicated in blood pressure control and in the pathogenesis of hypertension, cardiac hypertrophy, and heart failure. Thus, this gene product is a target for cardiovascular disease therapies. This gene maps to 14q11. 2 in a cluster of genes encoding other proteases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mast Cell Chymase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene CMA1
Gene Accession No. P21844, P23946, P50339
Gene Alias alpha-chymase; Chymase; chymase 1; chymase 1 preproprotein transcript E; chymase 1 preproprotein transcript I; chymase 1, mast cell; chymase, heart; chymase, mast cell; CMA1; CYH; CYM; mast cell; Mast cell chymase 1; mast cell protease 3; Mast cell protease 5; mast cell protease I; Mast cell protease III; MCP3P; Mcp5; Mcp-5; Mcpt3; Mcpt5; MCT1; mMCP-5; rMCP-3; rMCP-5; rMCP-III
Gene Symbols CMA1
Host Species Rabbit
Immunogen E.coli-derived human CMA1 recombinant protein (Position: I22-N247).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1215, 17228, 25627
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.