Learn More
CPC Scientific H-Phe-Leu-Pro-Leu-Ile-Leu-Gly-Lys-Leu-Val-Lys-Gly-Leu-Leu-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific MCDP002B
Encompass
MCD Peptide HR-1 is isolated from the venom of the giant hornet Vespa Orientalis and differs structurally from the MCD peptide from honeybee venom but possesses similar biological effects. These peptides are capable of degranulating rat mast cells, thereby initiating histamine release. ONE-LETTER SEQUENCE: FLPLILGKLVKGLL-NH2MOLECULAR FORMULA: C77H135N17O14MOLECULAR WEIGHT:1523.03STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [80388-04-1], SYNONYMS: Mast Cell Degranulating (MCD) Peptide HR-2RESEARCH AREA: InflammationREFERENCES: M.U. Tuichibaev et al., Biochem. USSR, 53, 183 (1988)
Catalog No. 50-210-9716
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.