Learn More
Greiner Bio-One MASTERBLOCK™ 1536 Deep Well Conical Bottom Storage Plate
GREENER_CHOICE
Catalog No. 07000013 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-000-013 Clear
1 options
Catalog No. 07-000-013 Supplier Greiner Bio-One Supplier No. 782270
Add to Cart
Add to Cart
Suitable for cultivating bacteria or yeasts. Greiner Bio-One MASTERBLOCK™ 1536 Deep Well Conical Bottom Storage Plate is also suitable for storing sample material.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Sealable with adhesive films and heat sealer
  • High temperature resistance
  • High chemical resistance
  • A
TRUSTED_SUSTAINABILITY
Specifications
Lid Without Lid
Array 32 x 48
Color Clear
Material Polypropylene
No. of Wells 1536
Sterility Non-sterile
Total Volume (Metric) Well 18 μL
Well Shape Rounded Square, Conical Bottom (V-well)
No. per Case 60
No. per Pack 15