Learn More
Greiner Bio-One MASTERBLOCK™ 384 Deep Well Polypropylene Conical Bottom Microplates
Catalog No. 07000048 Shop All Greiner Bio-One Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
07-000-048 Non-sterile
07-000-885 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 07-000-048 Supplier Greiner Bio-One Supplier No. 781270
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Free of detectable DNase, RNase, human DNA.
  • Alphanumeric well coding
  • High chemical resistanceHigh temperature resistanceSealable with adhesive films and heat sealer
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 16 x 24
Binding Type Non Binding
Color Clear
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 20 to 225 μL
Sterility Non-sterile
Total Volume (Metric) Well 240 μL
Well Shape Rounded Square, Conical Bottom (V-well)
No. per Case 60
No. per Pack 6
Quantity 60/Cs.
Well Color Clear