Learn More
Greiner Bio-One MASTERBLOCK™ 384 Deep Well Polypropylene Conical Bottom Microplates
Shop All Greiner Bio-One ProductsDescription
- Alphanumeric well coding
- High chemical resistance
- High temperature resistance
- Sealable with adhesive films and heat sealer
Service Plan
No
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Binding Type | Non Binding |
| Color | Clear |
| Material | Polypropylene |
| No. of Wells | 384 |
| Volume (Metric) Working | 20 to 225 μL |
| Sterility | Sterile |
| Total Volume (Metric) Well | 240 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.