Learn More
Greiner Bio-One MASTERBLOCK™ 96 Deep Well Round Bottom 1mL Storage Plate
Catalog No. 07000159 Shop All Greiner Bio-One Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
No. per Pack:
1
5
Bulk
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility No. per Pack
07-000-159 Sterile 1
07-000-118 Non-sterile 5
07-000-119 Non-sterile 1
07-000-120 Non-sterile Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07-000-159 Supplier Greiner Bio-One Supplier No. 780261
Add to Cart
Add to Cart
Suitable for cultivating bacteria or yeasts. Greiner Bio-One MASTERBLOCK™ 96 Deep Well Round Bottom 1mL Storage Plate is also suitable for storing sample material.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Sealable with adhesive films and heat sealer
  • High temperature resistance
  • High chemical resistance
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Color Natural
Material Polypropylene
No. of Wells 96
Sterility Sterile
Surface Treatment Untreated
Total Volume (Metric) Well 1 mL
Well Shape Round Bottom (U-well)
No. per Pack 1
Quantity 50/Cs.
Well Design Chimney