Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One MASTERBLOCK™ V-Bottom 1536-well Deep-well Storage Plate

Catalog No. 07000012 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-012 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-012 Supplier Greiner Bio-One Supplier No. 782279

missing translation for 'validMainframeMsg'

Non-sterile

TRUSTED_SUSTAINABILITY

Specifications

Array 32 x 48
Color Black
Material Polypropylene
No. of Wells 1536
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
For Use With (Application) Storage
No. per Case 60
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.