Learn More
StatLab MasterTech 3% Potassium Ferrocyanide

Description
- Used to stain for iron deposits in tissue samples specifically through a technique called Prussian blue staining where it reacts with ferrous iron to produce a blue colored precipitate allowing for visualization of iron-rich areas within the tissue
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| Concentration | 3% |
| Quantity | 100 mL |
| Product Type | Potassium Ferrocyanide |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.