Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Bluing Solution
SDP

Catalog No. 22021410
Change view
Click to view available options
Quantity:
1 pt.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-410 1 gal.
22-021-417 1 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-410 Supplier StatLab Supplier No. HXB00242E
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab MasterTech Bluing Solution - achieves precise cellular bluing every time

  • Solution can be used in a regressive or a progressive H and E staining procedure
  • Achieves precise cellular bluing every time
  • Used to change the color of hematoxylin-stained nuclei from red to blue

Specifications

Grade Chemical Solution
Product Type Bluing Reagent
Quantity 1 gal.
Solution Type Stain Kit
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.