Learn More
StatLab MasterTech Buffered Versenate
SDP
Catalog No. 21033359
Change view
Click to view available options
Solubility:
Buffered Decalcifier
Catalog No. Solubility
21-033-359 Buffered Decalcifier
1 options
Catalog No. 21-033-359 Supplier StatLab Supplier No. DCV0150
Add to Cart
Add to Cart
A non-aggressive chelating bone decalcifier
  • Compatible with all special stain and immunohistochemical techniques
  • This gentle bone decalcifier is pH balanced with a pH of 7.4, allowing all of the E.D.T.A. to be dissolved
  • A MasterTech Special Stain Kit Component
Specifications
Solubility Buffered Decalcifier