Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Diastase
SDP

Catalog No. 22021440
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-440 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-440 Supplier StatLab Supplier No. AHDIA50
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Diastase, a MasterTech Special Stain Kit Component

  • Diastase contains alpha-amylase and depolymerizes glycogen into smaller sugar units primarily maltose and glucose
  • These units are then removed from the tissue section by rinsing with water
  • Used to specifically identify and differentiate glycogen within a tissue sample by digesting it allowing for a comparison between a treated and untreated section when stained with Periodic Acid-Schiff (PAS) stain
  • A MasterTech Special Stain Kit Component

Specifications

Packaging Plastic Bottle
Sensitive to Heat Inactivation Yes
Quantity 50 g
Research Category Toxicology and Drug Metabolism, Drug Development
Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.