Learn More
StatLab MasterTech Eosin Y Stain
Description
- The addition of Phloxine B to this MasterTech Alcoholic Eosin formulation provides a richer warmer H and E counterstain
- A MasterTech Eosin Stain
- Ready-to-use
Specifications
Specifications
| Quantity | Pint |
| Product Type | Eosin Y Stain Phloxine B |