Learn More
StatLab MasterTech Harris Hematoxylin

Description
- Typically used as a regressive stain with MasterTech differentiating solution, this mercury free formulation is ideal for Histology applications
- Used in conjunction with Eosin for HandE staining to screen specimens in anatomic pathology, research, smears, touch preps and other applications
Specifications
Specifications
| For Use With (Equipment) | Staining Equipment |
| Grade | Chemical Solution |
| Product Type | Hematoxylin |
| Quantity | 1000 mL |
| Solution Type | Stain |
| Form | Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.